Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140264
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140264 Supplier Invitrogen™ Supplier No. PA5140264
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: APAPAKPKEE KIDLSAIKIE FSKEQQDEF Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 93%.

Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in fast skeletal muscle. Two transcript variants have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYL1
Gene Accession No. P05976
Gene Alias A1 catalytic; A2 catalytic; AI325107; alkali myosin light chain 1; alkali myosin light chain 3; alkali myosin light chain 3 A2 catalytic; Ca2+-binding protein-like protein; fast myosin alkali light chain 1; fast myosin alkali light chain 3; fast myosin light chain 1; fast myosin light chain 1F; Mcl3; MLC; Mlc1; MLC1/MLC3; MLC1F; MLC1F/MLC3F; Mlc3; MLC3F; MLC3-f; MLCf; Myl1; Mylf; myosin light chain; myosin light chain 1; myosin light chain 1, skeletal muscle isoform; myosin light chain 1/3, skeletal muscle isoform; myosin light chain 3; myosin light chain 3, skeletal muscle isoform; myosin light chain A1/A2; Myosin light chain alkali 1/2; myosin light chain, alkali, fast skeletal muscle; myosin, light chain 1; myosin, light chain 1, alkali; skeletal, fast; myosin, light polypeptide 1; myosin, light polypeptide 1, alkali; atrial, embryonic; myosin, light polypeptide 1, alkali; skeletal, fast; myosin, light polypeptide 4, alkali; atrial, embryonic
Gene Symbols MYL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYL1. Recombinant protein control fragment (Product #RP-109370).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4632
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.