Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYL10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571154
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Human: 100%; Mouse: 93%; Pig: 93%; Rat: 93%
MYL10 is a protein coding gene.
Specifications
MYL10 | |
Polyclonal | |
Unconjugated | |
MYL10 | |
1700027I08Rik; MYL10; Myl10-ps1; MYLC2PL; myosin light chain 10; myosin light chain 2, lymphocyte-specific; myosin light chain 2, precursor lymphocyte-specific; myosin regulatory light chain 10; myosin, light chain 10, regulatory; myosin, light chain 10, regulatory, pseudogene 1; PLRLC; Precursor lymphocyte-specific regulatory light chain; RGD1561258 | |
Rabbit | |
Affinity chromatography | |
RUO | |
93408 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BUA6 | |
MYL10 | |
synthetic peptide directed towards the N-terminal region of human MYL10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction