Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYL12A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597190 Supplier Invitrogen™ Supplier No. PA597190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLNGTDPEDV IRNAFACFDE EATGTIQEDY LRELLTTMGD RFTDEEVDEL YREAPIDKKG NFNYIEFTRI LKHGAKDKDD; Positive Samples: LO2, A375, HeLa, K-562, Jurkat.

This gene encodes a nonsarcomeric myosin regulatory light chain. This protein is activated by phosphorylation and regulates smooth muscle and non-muscle cell contraction. This protein may also be involved in DNA damage repair by sequestering the transcriptional regulator apoptosis-antagonizing transcription factor (AATF)/Che-1 which functions as a repressor of p53-driven apoptosis. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYL12A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Myl12a
Gene Accession No. P19105
Gene Alias 2900073G15Rik; AI838292; brain specific myosin regulatory light chain; C80200; Epididymis secretory protein Li 24; HEL-S-24; MLC-2; MLC20; MLC-2A; MLC-2B; MLCB; MRCL3; MRLC2; MRLC3; MYL12A; MYL12B; MYL2B; MYLC2B; myosin light chain 12A; myosin light chain, regulatory B-like; myosin regulatory light chain 12A; Myosin regulatory light chain 12B; Myosin regulatory light chain 2, nonsarcomeric; Myosin regulatory light chain 20 kDa; myosin regulatory light chain 2-A, smooth muscle isoform; Myosin regulatory light chain 2-B, smooth muscle isoform; myosin regulatory light chain 3; Myosin regulatory light chain MRLC2; Myosin regulatory light chain MRLC3; myosin regulatory light chain RLC-A; Myosin RLC; myosin RLC-A; myosin, light chain 12A, regulatory, non-sarcomeric; myosin, light polypeptide, regulatory, non-sarcomeric (20kD); NMDA receptor-interacting protein; RGD1309537; RLC; Rlc-a; SHUJUN-1
Gene Symbols Myl12a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 92-171 of human MYL12A (NP_0064621).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10627
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.