Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL2 (phospho Y118) Rabbit anti-Human, Polyclonal , Abnova™

Catalog No. 89956772 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-956-772 Supplier Abnova Corporation Supplier No. PAB29624
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MYL2.

Thus gene encodes the regulatory light chain associated with cardiac myosin beta (or slow) heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy. [provided by RefSeq]

Specifications

Antigen MYL2 (phospho Y118)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MYL2.
Dilution Immunofluorescence (1:100-1:200) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Gene Alias CMH10/DKFZp779C0562/MLC2
Gene Symbols MYL2
Host Species Rabbit
Immunogen A synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y118 of human MYL2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4633
Test Specificity MYL2 (phospho Y118) polyclonal antibody detects endogenous level of MYL2 only when phosphorylated at tyrosine 118.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.