Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578066
Description
Positive Control: human heart, Mouse heart, Rat heart Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.
Specifications
MYL2 | |
Polyclonal | |
Unconjugated | |
MYL2 | |
cardiac myosin light chain 2; cardiac myosin light chain-2; cardiac ventricular myosin light chain 2; CMH10; DKFZp779C0562; hypothetical protein LOC677750; Mlc2; MLC-2; mlc2b; MLC-2s/v; MLC2V; MLC-2v; myl2; myl2b; Mylpc; myosin light chain 2; myosin light chain 2, slow skeletal/ventricular muscle isoform; myosin light chain 2V; myosin light chain phosphatase; myosin light chain, phosphorylatable, cardiac ventricles; myosin light chain-2; myosin regulatory light chain 2, ventricular/cardiac muscle isoform; myosin regulatory light chain 2b; myosin regulatory light chain ventricular isoform; myosin, light chain 2, regulatory, cardiac, slow; myosin, light chain 2b, regulatory, cardiac, slow; myosin, light polypeptide 2, regulatory, cardiac, slow; myosin, light polypeptide 2b, regulatory, cardiac, slow; regulatory light chain of myosin; RLC of myosin; slow cardiac myosin regulatory light chain 2; ventricular myosin light chain 2; ventricular myosin regulatory light chain; zgc:136848 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17906, 363925, 4633 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.32 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P08733, P10916, P51667 | |
MYL2 | |
Recombinant protein encompassing a sequence within the center region of human Myosin Light Chain 2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction