Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587256
Description
Purity is > 95% by SDS-PAGE.
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.
Specifications
MYL2 | |
Polyclonal | |
Unconjugated | |
MYL2 | |
cardiac myosin light chain 2; cardiac myosin light chain-2; cardiac ventricular myosin light chain 2; CMH10; DKFZp779C0562; hypothetical protein LOC677750; Mlc2; MLC-2; mlc2b; MLC-2s/v; MLC2V; MLC-2v; myl2; myl2b; Mylpc; myosin light chain 2; myosin light chain 2, slow skeletal/ventricular muscle isoform; myosin light chain 2V; myosin light chain phosphatase; myosin light chain, phosphorylatable, cardiac ventricles; myosin light chain-2; myosin regulatory light chain 2, ventricular/cardiac muscle isoform; myosin regulatory light chain 2b; myosin regulatory light chain ventricular isoform; myosin, light chain 2, regulatory, cardiac, slow; myosin, light chain 2b, regulatory, cardiac, slow; myosin, light polypeptide 2, regulatory, cardiac, slow; myosin, light polypeptide 2b, regulatory, cardiac, slow; regulatory light chain of myosin; RLC of myosin; slow cardiac myosin regulatory light chain 2; ventricular myosin light chain 2; ventricular myosin regulatory light chain; zgc:136848 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17906, 363925, 4633 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P08733, P10916, P51667 | |
MYL2 | |
Recombinant full length Human Myosin light chain 2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction