Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYL3 Monoclonal Antibody (7C1)
GREENER_CHOICE

Catalog No. PIMA515514
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515514 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515514 Supplier Invitrogen™ Supplier No. MA515514
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15514 targets MYL3 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15514 immunogen is purified recombinant fragment of MYL3 expressed in E. Coli. MA5-15514 detects MYL3 which has a predicted molecular weight of approximately 22kDa.

MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYL3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7C1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene MYL3
Gene Accession No. P08590, P16409
Gene Alias Cardiac myosin light chain 1; CMH8; CMLC1; MLC1s; MLC1SB; Mlc1v; MLClV; MLC-lV/sb; MYL3; Mylc; Mylc1v; Myosin alkali light chain 1, ventricular; Myosin alkali light chain 1, ventricular/slow skeletal muscle isoform; Myosin light chain 1, slow-twitch muscle B/ventricular isoform; myosin light chain 3; Myosin light chain 3, alkali, cardiac ventricles; myosin light chain, alkali, cardiac ventricles; myosin, light chain 3, alkali; ventricular, skeletal, slow; myosin, light polypeptide 3; myosin, light polypeptide 3, alkali; ventricular, skeletal, slow; rVMLC1; slow skeletal; slow skeletal ventricular myosin alkali light chain 3; ventricular; ventricular myosin alkali light chain; Ventricular myosin light chain 1; ventricular, skeletal, slow; ventricular/slow twitch myosin alkali light chain; VLC1; VLCl
Gene Symbols MYL3
Host Species Mouse
Immunogen Purified recombinant fragment of MYL3 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24585, 4634
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.