Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYL3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2E4B2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | MYL3 |
---|---|
Clone | 2E4B2 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6718
![]() ![]() |
Proteintech Group Inc
662861IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy.Specifications
MYL3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Cardiac myosin light chain 1, CMH8, CMLC1, MLC1SB, MLC1V, MYL3, Myosin light chain 3, VLC1 | |
Myl3 | |
IgG1 | |
Protein G | |
Antibody |
2E4B2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
P08590, P09542, P16409 | |
100515755, 17897, 24585, 4634 | |
MYL3 Fusion Protein Ag24535 | |
Primary | |
-20°C | |
MYL3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title