Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL4 Antibody (4E8), Alexa Fluor™ 700, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242646AF700

Catalog No. NBP242646AI


Only null left
Add to Cart

Description

Description

MYL4 Monoclonal specifically detects MYL4 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

MYL4
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
MYL4
Recombinant human MYL4 (1-197aa) purified from E. coli
100 μL
Vision
4635
Store at 4C in the dark.
IgG2a κ
Western Blot, ELISA
400000000
Western Blot, ELISA
ALC1, AMLC, GT1, MLC1, Myosin light chain 1, embryonic muscle/atrial isoform, myosin light chain 4, Myosin light chain alkali GT-1 isoform, myosin, atrial/fetal muscle, light chain, myosin, light chain 4, alkali; atrial, embryonic, myosin, light polypeptide 4, alkali; atrial, embryonic, PRO1957
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.