Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYL4 Antibody (OTI1H6), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272856UV
Description
MYL4 Monoclonal specifically detects MYL4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MYL4 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MYL4 | |
Recombinant protein fragment corresponding to amino acids 2-197 of human MYL4 (NP_002467) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI1H6 | |
Western Blot, Immunohistochemistry | |
ALC1, AMLC, GT1, MLC1, Myosin light chain 1, embryonic muscle/atrial isoform, myosin light chain 4, Myosin light chain alkali GT-1 isoform, myosin, atrial/fetal muscle, light chain, myosin, light chain 4, alkali; atrial, embryonic, myosin, light polypeptide 4, alkali; atrial, embryonic, PRO1957 | |
Mouse | |
Affinity Purified | |
Vision | |
4635.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction