Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYL4 Monoclonal Antibody (OTI3H6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA807580

Description
Regulatory light chain of myosin. Does not bind calcium.Specifications
MYL4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P12829 | |
MYL4 | |
Human recombit protein fragment corresponding to amino acids 2-197 of human MYL4 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI3H6 | |
Unconjugated | |
MYL4 | |
ALC1, AMLC, GT1, PRO1957 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4635 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction