Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL4 Mouse anti-Human, Clone: OTI1H6, liquid, TrueMAB™
SDP

Catalog No. 501681627
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1627 Supplier OriGene Supplier No. TA807579
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Regulatory light chain of myosin. Does not bind calcium.

Specifications

Antigen MYL4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H6
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MYL4
Gene Accession No. P12829
Gene Alias ALC1, AMLC, GT1, PRO1957
Gene Symbols MYL4
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 2-197 of human MYL4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4635
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.