Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | MYL4 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8590
![]() ![]() |
Proteintech Group Inc
132291AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Regulatory light chain of myosin. Does not bind calcium.Specifications
MYL4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P09541, P12829, P17209 | |
17896, 4635, 688228 | |
MYL4 Fusion Protein Ag4031 | |
Primary | |
-20°C | |
MYL4 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ALC1, AMLC, GT1, MLC1, MYL4, Myosin light chain 4, PRO1957 | |
Myl4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title