Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYL5 Monoclonal Antibody (3D12)
GREENER_CHOICE

Catalog No. PIMA524578
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524578 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524578 Supplier Invitrogen™ Supplier No. MA524578
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: RKTKKKEGGA LRAQRASSNV FSNFEQTQIQ EFKEAFTLMD QNRDGFIDKE DLKDTYASLG KTNVKDDELD AMLKEASGPI NFTMFLNLFG EKL.

This gene encodes one of the myosin light chains, a component of the hexameric ATPase cellular motor protein myosin. Myosin is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene product, one of the regulatory light chains, is expressed in fetal muscle and in adult retina, cerebellum, and basal ganglia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYL5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D12
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MYL5
Gene Accession No. Q02045
Gene Alias MYL5; MYLC2; myLC-2; Myosin light chain 5; myosin regulatory light chain 5; myosin, light chain 5, regulatory; myosin, light polypeptide 5, regulatory; superfast myosin regulatory light chain 2
Gene Symbols MYL5
Host Species Mouse
Immunogen MYL5 (NP_002468, 4 a.a. ~ 96 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4636
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.