Learn More
Invitrogen™ MYL6 Polyclonal Antibody

Description
Immunogen sequence: MTEEEVEMLV AGHEDSNGCI NYEAF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 92%.
Specifications
Specifications
| Antigen | MYL6 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MYL6 |
| Gene Accession No. | P60660 |
| Gene Alias | 17 kDa myosin light chain; ESMLC; LC17; LC17A; LC17B; LC17-GI; LC17-NM; MLC1SM; MLC-3; MLC3NM; MLC3SM; Myl6; Myln; myosin light chain 3; myosin light chain 6; myosin light chain A3; Myosin light chain alkali 3; myosin light chain, alkali, nonmuscle; myosin light polypeptide 6; myosin, light chain 6, alkali, smooth muscle and non-muscle; myosin, light polypeptide 6, alkali, smooth muscle and non-muscle; smooth muscle and nonmuscle myosin light chain alkali 6 |
| Gene Symbols | MYL6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.