Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB123613 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB123613 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB123613 Supplier Novus Biologicals Supplier No. NBP309356100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MYL6 Polyclonal specifically detects MYL6 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MYL6
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Formulation PBS buffer, 2% sucrose
Gene Alias ESMLC, LC17, LC17A, LC17B, LC17-GI, LC17-NM, MLC1SM, MLC-3, MLC3NM, MLC3SM, Myosin light chain 3, Myosin light chain A3, Myosin light chain alkali 3, myosin light polypeptide 6,17 kDa myosin light chain, myosin, light chain 6, alkali, smooth muscle and non-muscle, myosin, light polypeptide 6, alkali, smooth muscle and non-muscle, Smooth muscle and nonmuscle myosin light chain alkali 6
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human MYL6 (NP_524147). Peptide sequence PMLQTVAKNKDQGTYEDYVEGLRVFDKEGNGTVMGAEIRHVLVTLGEKMT
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 4637
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.