Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYL6B Monoclonal Antibody (OTI7F10)
GREENER_CHOICE

Catalog No. PIMA527405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527405 Supplier Invitrogen™ Supplier No. MA527405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYL6B Monoclonal Antibody for Western Blot

Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in both slow-twitch skeletal muscle and in nonmuscle tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYL6B
Applications Western Blot
Classification Monoclonal
Clone OTI7F10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Myl6b
Gene Accession No. P14649, Q8CI43
Gene Alias 5730437E04Rik; BC037527; MLC1SA; MYL6B; myosin alkali light chain 1 slow a; myosin light chain 1 slow a; myosin light chain 1 slow-twitch muscle A isoform; myosin light chain 1, slow-twitch muscle A isoform; myosin light chain 6B; myosin, light chain 6B, alkali, smooth muscle and non-muscle; myosin, light polypeptide 6B; myosin, light polypeptide 6B, alkali, smooth muscle and non-muscle; sm; smooth muscle and non-muscle myosin alkali light chain 6B; smooth muscle and nonmuscle myosin light chain alkali 6B
Gene Symbols Myl6b
Host Species Mouse
Immunogen Full length human recombinant protein of MYL6B produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140465, 216459
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.