Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYL7 Antibody (OTI4E7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272859H
Description
MYL7 Monoclonal specifically detects MYL7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MYL7 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
MYL7 | |
Full length recombinant protein of human MYL7 (NP_067046) produced in E.coli. | |
0.1 mL | |
58498.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4E7 | |
Western Blot, Immunohistochemistry | |
atrial isoform, light polypeptide 7, regulatory, MLC-2a, MYL2A, MYLC2AMLC2a, myosin, light chain 7, regulatory | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction