Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL7 Monoclonal Antibody (OTI7F2), TrueMAB™, OriGene
SDP

Catalog No. 501678147
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-8147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-8147 Supplier OriGene Supplier No. TA800174
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Encoded by the MYL7 gene, myosin regulatory light chain 7, also designated Myosin regulatory light chain 2, atrial isoform (MLC-2a), is part of a hexameric complex of two heavy chains and four light chains predomitly expressed in adult atrial muscle. Myosin regulatory light chain 7 binds calcium and has been shown to be a useful molecular marker for cardiac chamber specification. The co-expression of myosin regulatory light chain 7 and myosin light chain 2 (MLC2v) in the outflow tract and atrioventricular canal, together with the single expression in the atrial (MYL7) and ventricular (MYL2) myo-cardium, permits the delineation of their boundaries. At the amino acid level there is 95% homology between the human and mouse myosin regulatory light chain 7 sequences.

Specifications

Antigen MYL7
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7F2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MYL7
Gene Accession No. Q01449
Gene Alias MYL2A, MYLC2A
Gene Symbols MYL7
Host Species Mouse
Immunogen Full length human recombit protein of human MYL7 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58498
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.