Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYL7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553041 Supplier Invitrogen™ Supplier No. PA553041
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DPEEAILSAF RMFDPSGKGV VNKDEFKQLL LTQADKFSPA EVEQMFALTP MDLAGNIDYK SLCYIITHGD EK Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.

Regulatory subunit of serine/threonine-protein phosphatase 4. May regulate the activity of PPP4C at centrosomal microtubule organizing centers. The PPP4C-PPP4R2-PPP4R3A PP4 complex specifically dephosphorylates H2AFX phosphorylated on 'Ser-140' (gamma-H2AFX) generated during DNA replication and required for DNA DSB repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYL7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Myl7
Gene Accession No. Q01449
Gene Alias MLC2a; MLC-2a; MLC-2alpha; MYL2A; MYL7; Mylc2a; myosin light chain 2a; myosin light chain 7; myosin light chain, regulatory A; myosin regulatory light chain 2, atrial isoform; Myosin regulatory light chain 7; myosin, light chain 7, regulatory; myosin, light polypeptide 7, regulatory; RLC-A
Gene Symbols Myl7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYL7. Recombinant protein control fragment (Product #RP-90718).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58498
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.