Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYL7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592314
Description
Immunogen sequence: MASRKAGTRG KVAATKQAQR GSSNVFSMFE QAQIQEFKEA FSCIDQNRDG IICKADLRET YSQLGKVSVP EEELDAMLQE GKGPINFTVF LTLFGEKLNG TDPEEAILSA FRMFDPSGKG VVNKDEFKQL LLTQADKFSP AEVEQMFALT PMDLAGNIDY KSLCYIITHG DEKEE; Positive Samples: Mouse lung, Mouse heart.
Regulatory subunit of serine/threonine-protein phosphatase 4. May regulate the activity of PPP4C at centrosomal microtubule organizing centers. The PPP4C-PPP4R2-PPP4R3A PP4 complex specifically dephosphorylates H2AFX phosphorylated on 'Ser-140' (gamma-H2AFX) generated during DNA replication and required for DNA DSB repair.
Specifications
MYL7 | |
Polyclonal | |
Unconjugated | |
Myl7 | |
MLC2a; MLC-2a; MLC-2alpha; MYL2A; MYL7; Mylc2a; myosin light chain 2a; myosin light chain 7; myosin light chain, regulatory A; myosin regulatory light chain 2, atrial isoform; Myosin regulatory light chain 7; myosin, light chain 7, regulatory; myosin, light polypeptide 7, regulatory; RLC-A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
17898 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.91 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9QVP4 | |
Myl7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-175 of human MYL7 (NP_0670461). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction