Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYL7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | MYL7 |
---|---|
Concentration | 0.36 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-559-955
![]() ![]() |
Proteintech Group Inc
172831AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Encoded by the MYL7 gene, myosin regulatory light chain 7, also designated Myosin regulatory light chain 2, atrial isoform (MLC-2a), is part of a hexameric complex of two heavy chains and four light chains predomitly expressed in adult atrial muscle. Myosin regulatory light chain 7 binds calcium and has been shown to be a useful molecular marker for cardiac chamber specification. The co-expression of myosin regulatory light chain 7 and myosin light chain 2 (MLC2v) in the outflow tract and atrioventricular canal, together with the single expression in the atrial (MYL7) and ventricular (MYL2) myo-cardium, permits the delineation of their boundaries. At the amino acid level there is 95% homology between the human and mouse myosin regulatory light chain 7 sequences.Specifications
MYL7 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q01449, Q9QVP4 | |
17898, 289759, 58498 | |
MYL7 Fusion Protein Ag11058 | |
Primary | |
-20°C | |
MYL7 |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MLC 2a, MLC2a, MYL2A, MYL7, MYLC2A, Myosin light chain 2a | |
Myl7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title