Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL9/MYL12, Rabbit anti-Human, Clone: F6, Abnova™

Catalog No. 89335594 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-594 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-594 Supplier Abnova Corporation Supplier No. MAB18760
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen MYL9/MYL12
Applications ELISA, Functional Assay, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone F6
Conjugate Unconjugated
Dilution ELISA Functional Study Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.1% BSA and 0.02% sodium azide).
Host Species Rabbit
Immunogen A peptide corresponding to N-terminus of MYL9.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.