Learn More
Invitrogen™ MYL9/MYL12 Recombinant Rabbit Monoclonal Antibody (AWBMyl9F6 (F-6))

Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Specifications
Specifications
| Antigen | MYL9/MYL12 |
| Applications | ELISA, Functional Assay, Immunohistochemistry |
| Classification | Recombinant Monoclonal |
| Clone | AWBMyl9F6 (F-6) |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MYL12B |
| Gene Accession No. | O14950, P19105, P24844, Q3THE2, Q9CQ19 |
| Gene Alias | 1500001M02Rik; 20 kDa myosin light chain; 2900073G15Rik; AI327049; AI838292; brain specific myosin regulatory light chain; C77744; C80200; Epididymis secretory protein Li 24; HEL-S-24; LC20; MLC2; MLC-2; MLC20; MLC-2A; MLC-2B; MLC-2C; MLCB; MLC-B; MRCL3; MRLC1; Mrlc2; MRLC3; Mrlcb; Myl12a; MYL12B; MYL2B; MYL9; MYLC2B; Mylc2c; myosin light chain 12A; myosin light chain 12B; myosin light chain 9; myosin light chain, regulatory B; myosin light chain, regulatory B-like; myosin light chain, regulatory C; myosin regulatory light chain 1; myosin regulatory light chain 12A; Myosin regulatory light chain 12B; myosin regulatory light chain 2; myosin regulatory light chain 2, nonsarcomeric; Myosin regulatory light chain 2, smooth muscle isoform; myosin regulatory light chain 20 kDa; myosin regulatory light chain 2-A, smooth muscle isoform; Myosin regulatory light chain 2-B, smooth muscle isoform; myosin regulatory light chain 3; myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; myosin regulatory light chain MRLC2; Myosin regulatory light chain MRLC3; myosin regulatory light chain RLC-A; myosin regulatory light chain-B; myosin regulatory light polypeptide 9; Myosin RLC; myosin RLC-A; Myosin RLC-B; myosin, light chain 12A, regulatory, non-sarcomeric; myosin, light chain 12B, regulatory; myosin, light chain 9, regulatory; myosin, light polypeptide 9, regulatory; myosin, light polypeptide, regulatory, non-sarcomeric (20kD); MYRL2; NMDA receptor-interacting protein; RGD1309537; RLC; Rlc-a; RLC-B; RLC-C; SHUJUN-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.