Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYL9 Polyclonal Antibody

Catalog No. PI720325
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI720325 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI720325 Supplier Invitrogen™ Supplier No. 720325

Rabbit Polyclonal Antibody

These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey and Cat.

Myosin, a structural component of muscle, consists of two heavy chains and four light chains. Myosin light chain may regulate muscle contraction by modulating the ATPase activity of myosin heads. It also binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYL9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4
Gene MYL9
Gene Accession No. P24844, Q9CQ19
Gene Alias 20 kDa myosin light chain; AI327049; DTNB; G1; Isoform L20-A; LC20; light polypeptide 9; MGC3505; MLC2; MLC-2; MLC20; MLC-2C; MRLC1; myl9; myl9.L; myl9b; myl9l; Mylc2c; myosin light chain 2; myosin light chain 9; myosin light chain 9 L homeolog; myosin light chain, regulatory C; myosin regulatory light chain; myosin regulatory light chain 1; myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 2, smooth muscle major isoform; myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; myosin regulatory light polypeptide 9; myosin regulatory light polypeptide 9b; myosin RLC; myosin, light chain 9, regulatory; myosin, light chain 9b, regulatory; myosin, light polypeptide 9, regulatory; Myrl2; RLC-C; smooth muscle major isoform; unnamed protein product; XELAEV_18003093mg; zgc:77916
Gene Symbols MYL9
Host Species Rabbit
Immunogen Peptides corresponding to Human MYL9 (aa 3-16, 111-124).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10398, 98932
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.