Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL9 Rabbit anti-Human, Mouse, Clone: 4C9E8, Novus Biologicals™
SDP

Catalog No. p-7248182 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168474 20 μg
NB168473 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168474 Supplier Novus Biologicals Supplier No. NBP31669120UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MYL9 Monoclonal antibody specifically detects MYL9 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen MYL9
Applications Western Blot
Classification Monoclonal
Clone 4C9E8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias LC2020 kDa myosin light chain, MLC2Myosin RLC, MRLC1MLC-2C, myosin regulatory light chain 1, Myosin regulatory light chain 2, smooth muscle isoform, Myosin regulatory light chain 9, Myosin regulatory light chain MRLC1, myosin regulatory light polypeptide 9, myosin, light chain 9, regulatory, myosin, light polypeptide 9, regulatory, MYRL2MGC3505
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MYL9 (P24844).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10398
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.