Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ MyLab™ Custom Kit - Exploring Gel Filtration Chromatography
SDPSCI_ED

Catalog No. S06468 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06468 9 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06468 Supplier EDVOTEK™ Supplier No. 1108

missing translation for 'validMainframeMsg'

Are you planning a virtual curriculum or looking for experiments ideal for distance learning? With the MyLab™ Custom Kits, you can easily plan your curriculum. Each kit comes with instructions and materials for 2-3 runs of each experiment.

Introduce chromatographic separation to your students! In this experiment, students separate dyes of different colors based on their size and shape. This experiment contains materials for dye separation which include dye sample, elution buffer and plastic disposables. Columns may be rinsed and reused.

  • Understand and practice chromatography
  • Explore how shape and size affect a molecule’s behavior
  • Separate colorful molecules using gel filtration chromatography
  • Includes instructions, dye samples, matrix, buffer, pipettes, microtiter plate, chromatography column

Specifications

Grade 9 to 15
Age 14 to 20
For Use With (Application) At Home Education
Includes Instructions, Dye Samples, Matrix, Buffer, Pipettes, Microtiter Plate, Chromatography Column
Quantity 1 Kit
Description Exploring Gel Filtration Chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.