Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ MyLab™ Custom Kit - Invisible Footprints
SDPSCI_ED

Catalog No. S06473 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
6 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06473 6 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06473 Supplier EDVOTEK™ Supplier No. 1930

missing translation for 'validMainframeMsg'

Are you planning a virtual curriculum or looking for experiments ideal for distance learning? With the MyLab™ Custom Kits, you can easily plan your curriculum. Each kit comes with instructions and materials for 2-3 runs of each experiment.

What would happen if each person was in charge of their own personal atmosphere? In this colorful experiment, students actually are! Explore the global carbon cycle and climate change in a simple, positive, and intimate way.

  • Learn about the carbon cycle and climate change
  • Examine, calculate, and emit a carbon footprint then see the effects
  • Create a positive carbon handprint and see its effects
  • Includes instructions, atmosphere and CO2 powder, color change solution, mitigation solution, tubes, pipettes

Specifications

Grade 6 to 15
Age 12 to 20
For Use With (Application) At Home Education
Includes Instructions, Atmosphere and CO2 Powder, Color Change solution, Mitigation Solution, Tubes, Pipettes
Quantity 1 Kit
Description Invisible Footprints
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.