Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYLIP Antibody (OTI3A2), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272862C
Description
MYLIP Monoclonal specifically detects MYLIP in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MYLIP | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MYLIP | |
Recombinant protein fragment corresponding to amino acids 127-385 of human MYLIP (NP_037394) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI3A2 | |
Western Blot | |
band 4.1 superfamily member BZF1, BZF1, cellular modulator of immune recognition (c-MIR), EC 6.3.2.-, Idol, IDOLmyosin regulatory light chain-interacting protein, Inducible degrader of the LDL-receptor, MIRE3 ubiquitin-protein ligase MYLIP, myosin regulatory light chain interacting proteinE3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism, Zinc Finger | |
29116.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction