Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYLIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596524 Supplier Invitrogen™ Supplier No. PA596524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RAITETHAFY RCDTVTSAVM MQYSRDLKGH LASLFLNENI NLGKKYVFDI KRTSKEVYDH ARRALYNAGV VDLVSRNNQS PSHSPLKSSE SSMNCSSCEG LSCQQTRVLQ EKLRKLKEAM LCMVCCEEEI NSTFCPCGHT VCCESCAAQL QSCPVCRSRV EHVQHVYLPT HTSLLNLTVI; Positive Samples: Jurkat, Mouse kidney, Mouse liver, Mouse spleen, Rat brain; Cellular Location: Cell membrane, Cytoplasm, Peripheral membrane protein.

The ERM protein family members ezrin, radixin, and moesin are cytoskeletal effector proteins linking actin to membrane-bound proteins at the cell surface. Myosin regulatory light chain interacting protein (MYLIP) is a novel ERM-like protein that interacts with myosin regulatory light chain and inhibits neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYLIP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MYLIP
Gene Accession No. D3ZDI6, Q8BM54, Q8WY64
Gene Alias 9430057C20Rik; band 4.1 superfamily member BZF1; BM-023; BZF1; cellular modulator of immune recognition (c-MIR); E3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor; E3 ubiquitin-protein ligase MYLIP; I79_001311; IDOL; inducible degrader of the LDL-receptor; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase MYLIP; MIR; Mylip; Myosin regulatory light chain interacting protein; myosin regulatory light chain-interacting protein; PP5242; RING-type E3 ubiquitin transferase MYLIP
Gene Symbols MYLIP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 266-445 of human MYLIP (NP_0373942).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218203, 29116, 306825
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.