Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYLIP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989250
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9250 Supplier Bioss Supplier No. BS9674R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The ERM protein family members ezrin, radixin, and moesin are cytoskeletal effector proteins linking actin to membrane-bound proteins at the cell surface. Myosin regulatory light chain interacting protein (MYLIP) is a novel ERM-like protein that interacts with myosin regulatory light chain and inhibits neurite outgrowth.

Specifications

Antigen MYLIP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MYLIP
Gene Accession No. Q8WY64
Gene Alias 9430057C20Rik; band 4.1 superfamily member BZF1; BM-023; BOS_21890; BZF1; cellular modulator of immune recognition (c-MIR); E3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor; E3 ubiquitin-protein ligase MYLIP; I79_001311; IDOL; Inducible degrader of the LDL-receptor; LOC100344866; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase MYLIP; MIR; MYLIP; Myosin regulatory light chain interacting protein; myosin regulatory light chain interacting protein-like; Myosin regulatory light chain-interacting protein; PP5242; RING-type E3 ubiquitin transferase MYLIP
Gene Symbols MYLIP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MYLIP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29116
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.