Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYLK2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP211574
Description
MYLK2 Polyclonal specifically detects MYLK2 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
| MYLK2 | |
| Polyclonal | |
| Western Blot 0.01 - 0.03 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| EC 2.7.11, EC 2.7.11.18, KMLC, MLCK2MLCK, myosin light chain kinase 2, myosin light chain kinase 2, skeletal/cardiac muscle, skeletal muscle, skMLCK | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize all reported isoforms (NP_444253.3; NP_444254.3 ; NP_444255.3 and NP_444256.3). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| MYLK2 | |
| Peptide with sequence C-KDTKNMEAKKLSKD corresponding to internal region according to NP_444253.3, NP_444254.3 NP_444255.3, NP_444256.3. | |
| 0.1 mg | |
| Protein Kinase | |
| 85366 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction