Learn More
Invitrogen™ MYLK2 Polyclonal Antibody

Description
Immunogen sequence: IEFQAVPSEK SEVGQALCLT AREEDCFQIL DDCPPPPAPF PHRMVELRTG NVSSEFSMNS KEALGGGKFG AVCTCMEKAT Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
Specifications
Specifications
| Antigen | MYLK2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MYLK2 |
| Gene Accession No. | Q9H1R3 |
| Gene Alias | 9830004H17Rik; BB138278; Klmc; KMLC; MLCK; MLCK2; Mylk2; myosin light chain kinase 2; myosin light chain kinase 2, skeletal muscle; myosin light chain kinase 2, skeletal/cardiac muscle; myosin, light polypeptide kinase 2; myosin, light polypeptide kinase 2, skeletal muscle; skMLCK |
| Gene Symbols | MYLK2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.