Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYLK3 Antibody [mFluor Violet 500 SE], Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP346974MFV500
Description
MYLK3 Polyclonal antibody specifically detects MYLK3 in Human samples. It is validated for ELISA,Immunohistochemistry,Western BlotSpecifications
MYLK3 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Western Blot | |
mFluor Violet 500 SE | |
caMLCK, Cardiac-MyBP-C-associated Ca/CaM kinase, EC 2.7.11, EC 2.7.11.18, MGC126319, MGC126320, MLC kinase, MLCK2, MLCKcardiac-MyBP-C associated Ca/CaM kinase, myosin light chain kinase 3, putative myosin light chain kinase 3 | |
myosin light chain kinase 3. (Uniprot# Q32MK0) | |
0.1 mL | |
Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
91807 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction