Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYLK4 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298058DL594
Description
MYLK4 Polyclonal specifically detects MYLK4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MYLK4 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| caMLCK like, EC 2.7.11, EC 2.7.11.1, myosin light chain kinase family, member 4, SGK085, SgK085myosin light chain kinase family member 4, Sugen kinase 85 | |
| Produced in rabbits immunized with E. coli-derived Human MYLK4 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 340156 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction