Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYLK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553393 Supplier Invitrogen™ Supplier No. PA553393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGLSPFLGDN DAETLNNILA CRWDLEDEEF QDISEEAKEF ISKLLIKEKS WRISASEALK HPWLSDHKLH SRLNAQKKKN RGSDAQDFV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 83%.

MYLK4 serine/threonine kinase or myosin light chain kinase family member 4 is a member of the CAMK kinase family. It is one of four myosin light chain kinases noted in a comprehensive analysis of protein kinases encoded by the human genome. MLCKs are activated by Ca2+-calmodulin complexes and require Mg2+ or Ca2+ as a co-factor. It has 2 isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYLK4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYLK4
Gene Accession No. Q86YV6
Gene Alias caMLCK like; MYLK4; Myosin light chain kinase family member 4; myosin light chain kinase family, member 4; SgK085; Sugen kinase 85
Gene Symbols MYLK4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYLK4. Recombinant protein control fragment (Product #RP-91446).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340156
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.