Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYLPF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103484
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103484 Supplier Invitrogen™ Supplier No. PA5103484
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MYLPF.

MYLPF (myosin light chain, phosphorylatable, fast skeletal muscle), also known as fast skeletal myosin light chain 2 or MLC2B, is a 169 amino acid protein that is expressed in fetal and adult skeletal muscle. A calcium binding protein, MYLPF contains three EF hand domains and is encoded by a gene that maps to human chromosome 16p11.2. Chromosome 16 encodes over 900 genes in approximately 90 million base pairs, makes up nearly 3% of human cellular DNA and is associated with a variety of genetic disorders. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYLPF
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MYLPF
Gene Accession No. P04466, P97457, Q96A32
Gene Alias 2410014J02Rik; DKFZp779C0757; DTNB; fast skeletal myosin light chain 2; G2; HUMMLC2B; MGC13450; MLC2; MLC-2; MLC2B; MLC2F; MRLC2; MYL11; Myl2; MYLPF; Myolc1; myosin light chain; myosin light chain 2; myosin light chain 2 type 2; myosin light chain 2 type I; myosin light chain, phosphorylatable, fast skeletal muscle; Myosin light polypeptide 2 alkali; myosin regulatory light chain 2, skeletal muscle isoform; Myosin regulatory light chain 2, skeletal muscle isoform type 2; myosin, light chain 11, regulatory; Myosin, light polypeptide 2, alkali; Myosin, light polypeptide 2, alkali; ventricular, skeletal, slow; phosphorylatable fast skeletal muscle myosin light chain; Unknown (protein for MGC:143425); ventricular skeletal slow; ventricular, skeletal, slow
Gene Symbols MYLPF
Host Species Rabbit
Immunogen A synthesized peptide derived from human MYLPF(Accession Q96A32), corresponding to amino acid residues T125-E169.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17907, 24584, 29895
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.