Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYLPF Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545808
Description
Peptide sequence: LEELLTTQCD RFSQEEIKNM WAAFPPDVGG NVDYKNICYV ITHGDAKDQE Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 77%.
MYLPF (myosin light chain, phosphorylatable, fast skeletal muscle), also known as fast skeletal myosin light chain 2 or MLC2B, is a 169 amino acid protein that is expressed in fetal and adult skeletal muscle. A calcium binding protein, MYLPF contains three EF hand domains and is encoded by a gene that maps to human chromosome 16p11.2. Chromosome 16 encodes over 900 genes in approximately 90 million base pairs, makes up nearly 3% of human cellular DNA and is associated with a variety of genetic disorders. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth.
Specifications
| MYLPF | |
| Polyclonal | |
| Unconjugated | |
| MYLPF | |
| 2410014J02Rik; DKFZp779C0757; DTNB; fast skeletal myosin light chain 2; G2; HUMMLC2B; MGC13450; MLC2; MLC-2; MLC2B; MLC2F; MRLC2; MYL11; Myl2; MYLPF; Myolc1; myosin light chain; myosin light chain 2; myosin light chain 2 type 2; myosin light chain 2 type I; myosin light chain, phosphorylatable, fast skeletal muscle; Myosin light polypeptide 2 alkali; myosin regulatory light chain 2, skeletal muscle isoform; Myosin regulatory light chain 2, skeletal muscle isoform type 2; myosin, light chain 11, regulatory; Myosin, light polypeptide 2, alkali; Myosin, light polypeptide 2, alkali; ventricular, skeletal, slow; phosphorylatable fast skeletal muscle myosin light chain; Unknown (protein for MGC:143425); ventricular skeletal slow; ventricular, skeletal, slow | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29895 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96A32 | |
| MYLPF | |
| Synthetic peptide directed towards the C-terminal of human MYLPF (aa 120-169). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction