Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYNN Monoclonal Antibody (OTI1C11), TrueMAB™, OriGene
SDP

Catalog No. 501678163
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-8163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-8163 Supplier OriGene Supplier No. TA800197
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a member of the BTB/POZ and zinc finger domain-containing protein family that are involved in the control of gene expression. Alternative splicing results in multiple transcript variants and a pseudogene has been identified on chromosome 14.

Specifications

Antigen MYNN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MYNN
Gene Accession No. Q9NPC7
Gene Alias OSZF, SBBIZ1, ZBTB31, ZNF902
Gene Symbols MYNN
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 10-300 of human MYNN produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55892
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.