Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYNN Monoclonal Antibody (OTI1F10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA800192

Description
This gene encodes a member of the BTB/POZ and zinc finger domain-containing protein family that are involved in the control of gene expression. Alternative splicing results in multiple transcript variants and a pseudogene has been identified on chromosome 14.Specifications
MYNN | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9NPC7 | |
MYNN | |
Human recombit protein fragment corresponding to amino acids 10-300 of human MYNN produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1F10 | |
Unconjugated | |
MYNN | |
OSZF, SBBIZ1, ZBTB31, ZNF902 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55892 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction