Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYNN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110002
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110002 Supplier Invitrogen™ Supplier No. PA5110002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQYSHHCEHL LERLNKQREA GFLCDCTIVI GEFQFKAHRN VLASFSEYFG AIYRSTSENN VFLDQSQVKA DGFQKLLEFI YTGTLNLDSW NVKEIHQAAD YLKVEEVVTK CKIKMEDFAF IANPSSTEIS SITGNIELNQ QTCLLTLRDY NNREKSEVST DLIQANPKQG ALAKKSSQTK KKKKAFNSPK TGQNKTVQYP SDILENASVE LFLDANKLPT PVVEQVAQIN DNSELELTSV VENTFPAQDI VHTVTVKRKR GKSQPNCALK EHSMSNIASV KSPYEAENSG EELDQRYSKA.

This gene encodes a member of the BTB/POZ and zinc finger domain-containing protein family that are involved in the control of gene expression. Alternative splicing results in multiple transcript variants and a pseudogene has been identified on chromosome 14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYNN
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MYNN
Gene Accession No. Q99MD8, Q9NPC7
Gene Alias 2810011C24Rik; AA415053; AI661031; AW049392; MYNN; Mynr; myoneurin; OSZF; SBBIZ1; ZBTB31; Zinc finger and BTB domain-containing protein 31; zinc finger protein with BTB/POZ domain; ZNF902
Gene Symbols MYNN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human MYNN (NP_061127.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55892, 80732
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.