Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYNN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530766 Supplier Invitrogen™ Supplier No. PA530766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (94%), Rat (81%), Xenopus laevis (81%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the BTB/POZ and zinc finger domain-containing protein family that are involved in the control of gene expression. Alternative splicing results in multiple transcript variants and a pseudogene has been identified on chromosome 14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYNN
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene MYNN
Gene Accession No. Q9NPC7
Gene Alias 2810011C24Rik; AA415053; AI661031; AW049392; MYNN; Mynr; myoneurin; OSZF; SBBIZ1; ZBTB31; Zinc finger and BTB domain-containing protein 31; zinc finger protein with BTB/POZ domain; ZNF902
Gene Symbols MYNN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Myoneurin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55892
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.