Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555019
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555019 Supplier Invitrogen™ Supplier No. PA555019
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRMKEQQELS LTEASLQKLQ ERRDQELRRL EEEACRAAQE FLESLNFDEI DECVRNIERS LSVGSEFSSE LAESACEEKP NFNFSQPYPE EEVDEGFEAD DDAFKDSPNP SEHGHSDQRT SGIRTSDDSS EEDPYMNDTV VPTSPSA Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

This gene encodes a member of the myosin superfamily. The protein represents an unconventional myosin; it should not be confused with the conventional non-muscle myosin-10 (MYH10). Unconventional myosins contain the basic domains of conventional myosins and are further distinguished from class members by their tail domains. This gene functions as an actin-based molecular motor and plays a role in integration of F-actin and microtubule cytoskeletons during meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO10
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYO10
Gene Accession No. Q9HD67
Gene Alias AW048724; D15Ertd600e; KIAA0799; mKIAA0799; MYO10; myosin X; myosin-X; myosin-X; unconventional myosin-X; unconventional myosin-10; Unconventional myosin-X; unconventionnal myosin-X
Gene Symbols MYO10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYO10. Recombinant protein control fragment (Product #RP-101077).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4651
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.