Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120163 Supplier Invitrogen™ Supplier No. PA5120163
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, A-431, BT-474, Mouse lung. The target is usually found in the following locations: Cell projection, Cytoplasm, Peripheral membrane protein, cell cortex, cytoskeleton, cytosol, filopodium membrane, filopodium tip, lamellipodium, ruffle. Immunogen sequence: EAELRAQQEE ETRKQQELEA LQKSQKEAEL TRELEKQKEN KQVEEILRLE KEIEDLQRMK EQQELSLTEA SLQKLQERRD QELRRLEEEA CRAAQEFLES.

This gene encodes a member of the myosin superfamily. The protein represents an unconventional myosin; it should not be confused with the conventional non-muscle myosin-10 (MYH10). Unconventional myosins contain the basic domains of conventional myosins and are further distinguished from class members by their tail domains. This gene functions as an actin-based molecular motor and plays a role in integration of F-actin and microtubule cytoskeletons during meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MYO10
Gene Accession No. F8VQB6, Q9HD67
Gene Alias AW048724; D15Ertd600e; KIAA0799; mKIAA0799; MYO10; myosin X; myosin-X; myosin-X; unconventional myosin-X; unconventional myosin-10; Unconventional myosin-X; unconventionnal myosin-X
Gene Symbols MYO10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 845-944 of human MYO10 (NP_036466.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17909, 4651
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.