Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO15A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585061
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585061 Supplier Invitrogen™ Supplier No. PA585061
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHPHLTRFLQ DVSRTPGLPF QGIAKACEQN LQKTLRFGGR LELPSSIELR AMLAGRSSKR QLFLLPGGLE RHLKIKTCTV ALDVVEEICA EMA Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 86%.

This gene encodes an unconventional myosin. This protein differs from other myosins in that it has a long N-terminal extension preceding the conserved motor domain. Studies in mice suggest that this protein is necessary for actin organization in the hair cells of the cochlea. Mutations in this gene have been associated with profound, congenital, neurosensory, nonsyndromal deafness. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Read-through transcripts containing an upstream gene and this gene have been identified, but they are not thought to encode a fusion protein. Several alternatively spliced transcript variants have been described, but their full length sequences have not been determined. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO15A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYO15A
Gene Accession No. Q9UKN7
Gene Alias DFNB3; MYO15; MYO15A; myosin XVA; myosin-XV; unconventional myosin-15; Unconventional myosin-XV
Gene Symbols MYO15A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYO15A. Recombinant protein control fragment (Product #RP-108704).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51168
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.