Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569176 Supplier Invitrogen™ Supplier No. PA569176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rat: 86%.

Myosins are actin-based motor molecules with ATPase activity. Unconventional myosins serve in intracellular movements. Their highly divergent tails are presumed to bind to membranous compartments, which would be moved relative to actin filaments. May be involved in targeting of the catalytic subunit of protein phosphatase 1 during brain development. Activates PI3K and concomitantly recruits the WAVE1 complex to the close vicinity of PI3K and regulates neuronal morphogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO16
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Myo16
Gene Accession No. Q9Y6X6
Gene Alias BM140241; C230040D10Rik; Gm1105; Kiaa0865; LOW QUALITY PROTEIN: unconventional myosin-XVI; mKIAA0865; MYAP3; MYO16; myo16 {ECO:0000250; MYO16 variant protein; Myo16b; Myosin heavy chain myr 8; myosin XVI; myosin-XVI; myosin-XVI; unconventional myosin-XVI; Myr8; neuronal tyrosine phosphorylated adaptor for phosphoinositide 3-kinase adaptor 3; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adapter 3; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 3; NYAP3; PPP1R107; protein phosphatase 1, regulatory subunit 107; unconventional myosin-16; Unconventional myosin-XVI; UniProtKB:Q9Y6X6}
Gene Symbols Myo16
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human MYO16.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23026
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.