Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYO16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103941
Description
Antibody detects endogenous levels of total MYO16.
Myosins are actin-based motor molecules with ATPase activity. Unconventional myosins serve in intracellular movements. Their highly divergent tails are presumed to bind to membranous compartments, which would be moved relative to actin filaments. May be involved in targeting of the catalytic subunit of protein phosphatase 1 during brain development. Activates PI3K and concomitantly recruits the WAVE1 complex to the close vicinity of PI3K and regulates neuronal morphogenesis.
Specifications
MYO16 | |
Polyclonal | |
Unconjugated | |
Myo16 | |
BM140241; C230040D10Rik; Gm1105; Kiaa0865; LOW QUALITY PROTEIN: unconventional myosin-XVI; mKIAA0865; MYAP3; MYO16; myo16 {ECO:0000250; MYO16 variant protein; Myo16b; Myosin heavy chain myr 8; myosin XVI; myosin-XVI; myosin-XVI; unconventional myosin-XVI; Myr8; neuronal tyrosine phosphorylated adaptor for phosphoinositide 3-kinase adaptor 3; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adapter 3; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 3; NYAP3; PPP1R107; protein phosphatase 1, regulatory subunit 107; unconventional myosin-16; Unconventional myosin-XVI; UniProtKB:Q9Y6X6} | |
Rabbit | |
Affinity chromatography | |
RUO | |
192253, 23026, 244281 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5DU14, Q9ERC1, Q9Y6X6 | |
Myo16 | |
A synthesized peptide derived from human MYO16(Accession Q9Y6X6), corresponding to amino acid residues Y1076-G1126. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction