Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYO18A Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079840AF750
Description
MYO18A Polyclonal antibody specifically detects MYO18A in Human samples. It is validated for Western BlotSpecifications
MYO18A | |
Polyclonal | |
Western Blot | |
KIAA0216DKFZp686L0243, MAJN, Molecule associated with JAK3 N-terminus, myosin 18A, Myosin containing a PDZ domain, myosin containing PDZ domain, myosin XVIIIA, myosin-XVIIIa, MYSPDZ, SP-A receptor subunit SP-R210 alphaS, SPR210 | |
The immunogen recognized by this antibody maps to a region between residue 2004 andd 2054 of human myosin 18A using the numbering given in entry NP_510880.2 (GeneID 399687). | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
399687 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction