Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO18A Monoclonal Antibody (OTI3F7), TrueMAB™, OriGene
SDP

Catalog No. 502543760
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3760 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3760 Supplier OriGene Supplier No. CF812216
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYO18A Monoclonal antibody specifically detects MYO18A in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MYO18A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3F7
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene BMP6
Gene Accession No. P20722, P22004, Q04906
Gene Alias CD245; KIAA0216; MAJN; molecule associated with JAK3 N-terminus; Myo18a; MyoPDZ; myosin 18a; myosin containing a PDZ domain; myosin containing PDZ domain; myosin XVIIIa; myosin XVIIIb; myosin-XVIIIa; MysPDZ; SP-A receptor subunit SP-R210 alphaS; SPR210; SP-R210; Surfactant protein receptor SP-R210; Unconventional myosin-XVIIIa
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 21-374 of human BMP6 (NP_001709) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12161, 25644, 654
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.