Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO18A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA554041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554041 Supplier Thermo Scientific Supplier No. PA554041

Rabbit Polyclonal Antibody

Immunogen sequence: MEVMEMEVME ARLIRAAEIN GEVDDDDAGG EWRLKYERAV REVDFTKKRL QQEFEDKLEV EQQNKRQLER RLGDLQADSE ESQRALQQLK KKCQRLT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

MYO18A may link Golgi membranes to the cytoskeleton and participate in the tensile force required for vesicle budding from the Golgi. Thereby, may play a role in Golgi membrane trafficking and could indirectly give its flattened shape to the Golgi apparatus. Alternatively, in concert with LURAP1 and CDC42BPA/CDC42BPB, has been involved in modulating lamellar actomyosin retrograde flow that is crucial to cell protrusion and migration. May be involved in the maintenance of the stromal cell architectures required for cell to cell contact. Regulates trafficking, expression, and activation of innate immune receptors on macrophages. Plays a role to suppress inflammatory responsiveness of macrophages via a mechanism that modulates CD14 trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO18A
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYO18A
Gene Accession No. Q92614
Gene Alias CD245; KIAA0216; MAJN; molecule associated with JAK3 N-terminus; Myo18a; MyoPDZ; myosin 18a; myosin containing a PDZ domain; myosin containing PDZ domain; myosin XVIIIa; myosin XVIIIb; myosin-XVIIIa; MysPDZ; SP-A receptor subunit SP-R210 alphaS; SPR210; SP-R210; Surfactant protein receptor SP-R210; Unconventional myosin-XVIIIa
Gene Symbols MYO18A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYO18A
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399687
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.